missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 130 mm Box Height

Product Code. 12439530 Shop All Tenak Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12439530

Brand: Tenak™ TE23336

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel

Specifications

Array 2 x 5
Holds Boxes
Material Stainless Steel
Positions 10
For Use With (Equipment) Upright Freezers
Dimensions (L x W x H) 683 x 140 x 268 mm
Height (Metric) 130 mm (Box)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.