missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 65 mm Shelf Height

Product Code. 15240904 Shop All Tenak Products
Change view
Click to view available options
Holds:
Microtiterplates
Unit Size:
Each
Product Code. Holds unitSize
15240904 Microtiterplates Each
15250904 Microtiterplates Each
15200904 Microtiterplates Each
15210904 Microtiterplates Each
15220904 Microtiterplates Each
15230904 Microtiterplates Each
15280894 Microtiterplates Each
15290894 Microtiterplates Each
8 options
This item is not returnable. View return policy

Product Code. 15240904

Brand: Tenak™ TE27202

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.

  • For use in chest freezers
  • Racks come with a locking rod to secure the cryoboxes
  • Constructed of stainless steel

Specifications

Holds Microtiterplates
Material Stainless Steel
Positions 10
For Use With (Equipment) Chest Frseezers
Dimensions (L x W x H) 135 x 92 x 669 mm
Includes Locking rod
Height (Metric) 65 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.