Learn More
METTLER TOLEDO Standard MA Precision Balance
Product Code. 18092364 Shop All Mettler Toledo Products
Change view
Click to view available options
Capacity:
120 g
220 g
320 g
520 g
620 g
1200 g
2200 g
3.2 kg
4.2 kg
5.2 kg
6.2 kg
12.2 kg
16.2 kg
32.2 kg
Readability:
1 mg
10 mg
0.1 g
1 g
Model:
MA1002
MA103
MA12001L
MA16001L
MA2002
MA203
MA3002
MA303
MA32000L
MA32001L
MA4002
MA5001
MA503
MA6002
MA602
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Readability Model unitSize
18092364 520 g 1 mg MA503 Each
18022314 120 g 1 mg MA103 Each
18052314 220 g 1 mg MA203 Each
18072364 320 g 1 mg MA303 Each
18002324 620 g 10 mg MA602 Each
18082314 1200 g 10 mg MA1002 Each
18012264 2200 g 10 mg MA2002 Each
18032264 3.2 kg 10 mg MA3002 Each
18042374 4.2 kg 10 mg MA4002 Each
18022324 6.2 kg 10 mg MA6002 Each
18052324 5.2 kg 0.1 g MA5001 Each
18042324 12.2 kg 0.1 g MA12001L Each
18082264 16.2 kg 0.1 g MA16001L Each
18012334 32.2 kg 0.1 g MA32001L Each
18052334 32.2 kg 1 g MA32000L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 18092364 Supplier METTLER TOLEDO Supplier No. 30697429
METTLER TOLEDO Standard MA Precision Balances are highly accurate, simple to operate and built with high quality materials for proven long lasting performance.
The precise weigh cell and internal adjustment provide accurate results. Intuitive onscreen guidance within the LCD hybrid touchscreen allows you to easily navigate through 10 built in applications and manage weighing data. The metal base and overload protection defend the balance against rough handling in any weighing environment.
MA Precision Balances include:
  • Internal adjustment
  • LCD hybrid touchscreen
  • 10 built-in applications
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The MA Balance is the successor to the ME-T Balance.See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, RS232
Capacity 520 g
Display Type Touchscreen
Readability 1 mg
Material Metal, PBT
Model MA503
Stabilization Time 1.5 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No