missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO Standard MA-P Precision Balance

Product Code. 18022274 Shop All Mettler Toledo Products
Click to view available options
Capacity:
620 g
2200 g
6.2 kg
Readability:
10 mg
0.1 g
Model:
MA2002P
MA6001P
MA602P
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18022274

Brand: METTLER TOLEDO 30697493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

METTLER TOLEDO Standard MA-P Precision Balances are simple to operate and portable for on-the-go weighing, without compromising on accuracy or performance.

The precise weigh cell delivers highly accurate results with each use. Increase efficiency in your workflow with the intuitive touchscreen that guides you through each weighing procedure. Easily export data to a connected printer, computer, or USB device.

MA-P Balances include:

  • Strain gauge weighing cell
  • Internal adjustment
  • LCD hybrid touchscreen
  • 10 built-in applications
  • Power cable and battery operated
  • Front level indicator
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option

The MA Compact is the successor to the PL-E Balance. See how it ranks among our balance lines:

  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY

Specifications

Connection Type USB-A, RS232
Capacity 6.2 kg
Display Type Touchscreen
Readability 0.1 g
Material PBT
Model MA6001P
Stabilization Time 1 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz, or 8 AA Batteries
Legal for Trade No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.