Learn More
METTLER TOLEDO Standard LA Precision Balance
Product Code. 18082274 Shop All Mettler Toledo Products
Change view
Click to view available options
Capacity:
220 g
420 g
2200 g
4200 g
Readability:
1 mg
10 mg
Model:
LA2002E
LA203E
LA4002E
LA403E
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Readability Model unitSize
18082274 420 g 1 mg LA403E Each
18012344 4200 g 10 mg LA4002E Each
18042384 220 g 1 mg LA203E Each
18002284 2200 g 10 mg LA2002E Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18082274 Supplier METTLER TOLEDO Supplier No. 30823722
METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.
Increase efficiency in your daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.
LA Precision Balances include:
  • Adjustment with external weights
  • Bright backlit LCD Display
  • 3 built-in weighing applications
  • Front level indicator
  • Overload protection
  • Metal base, ABS top housing
  • RS232 interface
  • Bluetooth option
  • Transfer weight function for easy data transfer
See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, RS232
Capacity 420 g
Display Type Backlit LCD
Readability 1 mg
Material Aluminum, ABS
Model LA403E
Stabilization Time 3 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No