Learn More
METTLER TOLEDO Standard LA Precision Balance
Product Code. 18002284 Shop All Mettler Toledo Products
Click to view available options
Capacity:
220 g
420 g
2200 g
4200 g
Readability:
1 mg
10 mg
Model:
LA2002E
LA203E
LA4002E
LA403E
Unit Size:
Each
Product Code. 18002284
Brand: METTLER TOLEDO 30853155
This item requires a price update prior to being ordered.
Request a Price for this item here.
METTLER TOLEDO Standard LA Precision Balances are cost efficient, accurate and designed for long lasting performance. These models offer simple weighing and have external adjustment.
Increase efficiency in your daily weighing operations with the bright, backlit LCD display and 3 built-in applications. Robust construction with a metal base and overload protection defend the balance against rough handling and tough weighing environments.
LA Precision Balances include:
  • Adjustment with external weights
  • Bright backlit LCD Display
  • 3 built-in weighing applications
  • Front level indicator
  • Overload protection
  • Metal base, ABS top housing
  • RS232 interface
  • Bluetooth option
  • Transfer weight function for easy data transfer
See how it ranks among our balance lines:
  • XPR Excellence Precision Balances
  • MX Advanced Precision Balances
  • MR Advanced Precision Balances
  • MA Standard Precision Balances
  • ME Standard Precision Balances
  • LA Standard Precision Balances
TRUSTED_SUSTAINABILITY
Specifications
Connection Type USB-A, RS232
Capacity 2200 g
Display Type Backlit LCD
Readability 10 mg
Material Aluminum, ABS
Model LA2002E
Stabilization Time 3 sec.
Type Precision Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No