Learn More
Koettermann™ Standard Hanging Cabinet, 600x480x366mm
Koettermann™ Standard Hanging Cabinet, 600x480x366mm provides suitable storage to accommodate a variety of laboratory needs.
Brand: Koettermann™ 307-00185
Additional Details : Weight : 18.40000kg
Description
- Wall-mounted cabinet
- One door
- One shelf
- Side panels
Specifications
366 x 600 x 480 mm | |
Left Swinging Door | |
No |
Wall Mounted Cabinet | |
1 | |
1 |