Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Adaptor Set, BioTitan retention, 1/4 in. to 3/8 in. (1/16 in. wall)
Product Code. 32178698 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 set
Unit Size:
Pack of 25
Product Code. Quantity Unit Size
32178698 1 set Pack of 25
1 options
Product Code. 32178698 Supplier Thermo Scientific™ Supplier No. SH31277.01
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific Standard Fluid Transfer Assembly with BioTitan products increase single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermenters right where you need them.
Thermo Scientific Standard Fluid Transfer Assembly with BioTitan products increase single-use product flexibility. They connect to our standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermenters right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Now manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 1/4 to 3/8 in.
For Use With (Application) Single-Use Bioprocessing
Quantity 1 set
Product Type Adaptor Set