Learn More
Sherwood Scientific™ Standard Cleaning Solution
Product Code. 12971821
Click to view available options
Type:
Standard Cleaning Solution
Unit Size:
500mL
Product Code. 12971821
Brand: Sherwood Scientific™ 00156184
Specifications
Type Standard Cleaning Solution
For Use With (Equipment) Flame photometers, chloride meters, and colorimeters
Quantity 500 mL