missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution pH 1.68, SI Analytics™

Product Code. 10472101 Shop All SI Analytics Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. Quantity unitSize
10472101 250 mL 250mL
1 options
This item is not returnable. View return policy

Product Code. 10472101

Brand: SI Analytics 285137677

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 1.68 resists changes in pH when small quantities of an acid are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, potassium tetraoxalate dihydrate
  • Store tigthly sealed at temperatures between 15°C and 25°C
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Rinse off of bigger amounts into drains or the aquatic environment may lead to decreased pH-values. A low pH-value harms aquatic organisms. In the dilution of the use-level the pH-value is considerably increased, so that after the use of the product the aqueous waste, emptied into drains, is only low water-dangerous
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Density 1.01g/cm3
pH 1.68
Physical Form Liquid
Quantity 250 mL
Formulation Water, potassium tetraoxalate dihydrate
Odor Odorless
Packaging DURAN™ Bottle
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.