missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Standard Benchtop Chilling/Heating Block

Product Code. 11860243
Click to view available options
Length (Metric):
46.4 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Compact, economical benchtop chilling/heating block. Cole-Parmer™ Standard Benchtop Chilling/Heating Block features an nergy-efficient Peltier design that chills or heats biological samples from -10 to 100°C with ±1°C digital control.

  • Chill, freeze, heat, or mix biological samples at your work station
  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes and plates, and more
  • Two-line LCD that reads actual and set point temperatures
  • 30-day countdown timer with alarm and auto/off
  • Built-in data logger, and RS-232 I/O port for bidirectional communication
  • Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
  • Ideal for a variety of applications such as storing enzymes or DNA libraries, maintaining 14°C for ligation reactions, and incubating or deactivating enzyme reactions.

Specifications

For Use With (Equipment) For Centrifuge Tubes, Assay Plates, PCR Tubes & Plates
Material Aluminum
Temperature Range (Metric) -10°C to +100°C
Type Refrigerated Incubator
Includes Chiller/Heater Module, Power Cord, Instruction Manual, and a 12 VDC Power Supply
Temperature Stability ±1°C
Dimensions (L x W x H) 46.4 x 42.8 x 37.4 mm
Length (Metric) 46.4 mm
Width (Metric) 42.8 mm
Height (Metric) 37.4 mm
Voltage 100 to 230 VAC
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'