Learn More
KERN and SOHN Stand BFS
Click to view available options
Type:
Scale Stand
Description
- Tripod for raising the display unit to a height of approximately 750mm
- Height of stand approx. 1040 mm
- Suitable for weighing beams KERN UFA, floor scales KERN BFB, KERN BFN, and KERN BFS, pallet scales KERN UFB and KERN UFN, and drive-through scale KERN NFB
Specifications
Specifications
| For Use With (Equipment) | KERN Scales |
| Type | Scale Stand |