missing translation for 'onlineSavingsMsg'
Learn More

STAMP2/STEAP4 Antibody (418714) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB4626JF635

Product Code. 30171496

  • £442.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

STAMP2/STEAP4 Monoclonal antibody specifically detects STAMP2/STEAP4 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STAMP2/STEAP4
Monoclonal
Janelia Fluor 635
EC 1.16.1, EC 1.16.1.-, FLJ23153, Six-transmembrane epithelial antigen of prostate 4, SixTransMembrane protein of prostate 2, STAMP2DKFZp666D049, STEAP family member 4, TIARPsix transmembrane prostate protein 2, TNFAIP9, Tumor necrosis factor, alpha-induced protein 9metalloreductase STEAP4, tumor necrosis-alpha-induced adipose-related protein
NS0 mouse myeloma cell line transfected with human STAMP2, Met1-His459, Accession # EAW76907
RUO
Primary
Human
Liquid
Flow Cytometry, CyTOF
418714
50mM Sodium Borate
Mouse
0.1 mL
Cancer
79689
Store at 4°C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.