missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAMBPL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | STAMBPL1 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16825565
|
Proteintech
27315-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16815565
|
Proteintech
27315-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAMBPL1 is zinc metalloprotease that specifically cleaves ′Lys-63′-linked polyubiquitin chains. Does not cleave ′Lys-48′-linked polyubiquitin chains.Specifications
| STAMBPL1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96FJ0 | |
| 57559 | |
| STAMBPL1 Fusion Protein Ag26308 | |
| Primary | |
| -20°C | |
| STAMBPL1 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ALMalpha, AMSH FP, AMSH like protease, AMSH LP, AMSHLP, KIAA1373, STAM binding protein like 1, STAMBPL1 | |
| STAMBPL1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title