missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAMBP Monoclonal Antibody (K56003_18H5), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555748

 View more versions of this product

Product Code. 30202242

  • £416.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: KSAVIPEKKD TVKKLKEIAF PKAEELKAEL LKRYTKEYTE YNEEKKKEAE ELARNMAIQQ ELEKEKQRVA QQKQQQLEQE QFHAFEEMIR NQELEKERLK IVQEFGKVDP GLGGPLVPDL EKPSLDVFPT LTVSSIQPSD CHTTVRPAKP PVVDRSLKPG ALSNSESIPT

Cytokine-mediated signal transduction in the JAK-STAT cascade requires the involvement of adaptor molecules. One such signal-transducing adaptor molecule contains an SH3 domain that is required for induction of MYC and cell growth. The protein encoded by this gene binds to the SH3 domain of the signal-transducing adaptor molecule, and plays a critical role in cytokine-mediated signaling for MYC induction and cell cycle progression. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STAMBP
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O95630
Stambp
Recombinant human STAMBP (immunogen range is 81-250).
50 μL
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot
K56003_18H5
Unconjugated
Stambp
5330424L14Rik; 5730422L11Rik; AMSH; associated molecule with the SH3 domain of STAM; AW107289; Endosome-associated ubiquitin isopeptidase; MGC126516; MGC126518; MICCAP; mKIAA4198; Stam binding protein; STAM-binding protein; Stambp; testicular secretory protein Li 54
Mouse
Protein A
RUO
10617
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.