missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13009-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation.Specifications
| STAM2 | |
| Polyclonal | |
| Unconjugated | |
| STAM2 | |
| Hbp, Hrs binding protein, STAM 2, STAM2, STAM2A, STAM2B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10254, 311030, 56324 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75886, O88811, Q5XHY7 | |
| Stam2 | |
| STAM2 Fusion Protein Ag3698 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction