missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAM Recombinant Rabbit Monoclonal Antibody (HL1637)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556886
This item is not returnable.
View return policy
Description
STAM Recombinant Monoclonal Antibody for Western Blot, ICC/IF
STAM (Signal transducing adaptor molecule) is an adaptor molecule involved in downstream signaling pathways mediated by growth factors and cytokines. STAM also plays an important role in endocytosis, the JAK-STAT signaling pathway, and membrane trafficking. In humans, the gene encoding STAM is located on chromosome 10.
Specifications
| STAM | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P70297, Q92783 | |
| STAM | |
| Recombinant fragment of human STAM. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1637 | |
| Unconjugated | |
| STAM | |
| DKFZp686J2352; HSE1 homolog; RGD1564499; signal transducing adapter molecule 1; signal transducing adaptor molecule; signal transducing adaptor molecule (SH3 domain and ITAM motif) 1; Stam; STAM1; STAM-1; STAT91 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20844, 8027 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction