missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAM-1 Antibody (29C678), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56533AF647
This item is not returnable.
View return policy
Description
STAM-1 Monoclonal specifically detects STAM-1 in Human samples. It is validated for Western Blot.
Specifications
| STAM-1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8027 | |
| Human | |
| Purified |
| Western Blot | |
| 29C678 | |
| Western Blot | |
| Q92783 | |
| STAM | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 149-165 of human STAM. | |
| 0.1 mL | |
| Primary | |
| A 59 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction