Learn More
Invitrogen™ eBioscience™ StainTray™
Product Code. 15518996
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity Unit Size
15518996 1 each Each
1 options
Product Code. 15518996 Supplier Invitrogen™ Supplier No. 44040410
  • ONLINE EXCLUSIVE PRICE
    £304.00
    £267.65 / Pack of 1
    Save £36.35
    11% Off
View all promotions
Estimated Shipment: 24-11-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
May be used as a humidified, light-protected chamber for staining slides for immunohistochemistry or immunocytochemistry. The StainTray™ has a black base made of tough ABS plastic withstanding a wide range of chemicals (avoid chlorinated hydrocarbons).
  • Accepts up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold slides even if the tray is held at an angle
  • When humidity is needed, wells between rails will hold up to 1mL of water securely without splashing
  • Middle wells will hold up to 2mL each
  • Rails are raised not only to avoid water touching the slides but also to make slides more easily retrievable
  • The base will also hold excess stain solution dripping from the slides
  • Four rubber feet ensure greater base stability
  • Units are stackable for storage and space saving purposes
  • A black lid is included for work with fluorescent reagents. It is made of ABS and can toleratae a temperature range of −80°C to 80°C
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stain Tray
Slide Capacity 20
Product Line eBioscience
For Use With (Application) Immunocytochemistry, Immunohistochemical Staining, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Microscopy
Quantity 1 each