Learn More
Erlab Stainless Steel Worksurface Captair 483 Smart
Product Code. 15545164 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Captair 483 Ductless Filtering Fume Hood
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15545164 Captair 483 Ductless Filtering Fume Hood Each
1 options
Product Code. 15545164 Supplier Erlab Supplier No. I0164050001
  • ONLINE EXCLUSIVE PRICE
    £1785.00
    £1713.48 / Pack of 1
    Save £71.52
    4% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Worksurface for your ductless fume hoods.
  • Worksurface for your ductless fume hood 483 Smart
  • Material: Stainless steel
  • Equipment: Spill rentention tray
Specifications
For Use With (Equipment) Captair 483 Ductless Filtering Fume Hood
Product Type Work Surface