Learn More
Labconco™ Stainless Steel Work Surface for XPert™ and RXPert™ Units
Product Code. 31934196 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Stainless Steel (304)
Unit Size:
Each
Product Code. Material Unit Size
31934196 Stainless Steel (304) Each
1 options
Product Code. 31934196 Supplier Labconco™ Supplier No. 3975504
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel work surface designed for XPert and RXPert units.
  • Type 304 Stainless Steel work surfaces have a 1 in. deep front spill trough and fit the dimensions of XPert Systems and Stations.
  • 4 ft. width
Specifications
Product Type Work Surface
Color Silver
No. of Doors None
No. of Drawers None
Material Stainless Steel (304)