missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Wire Shelf

Product Code. 12893486
Click to view available options
For Use With (Equipment):
IB-15G
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12893486

Brand: Medline Scientific™ EDA8135

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Material: Stainless Steel; For Use With: IB-15G

  • Electro-polished
  • Available in a range of sizes and counts
  • For use with Medline Scientific™ incubators

Specifications

For Use With (Equipment) IB-15G
Material Stainless Steel
No. of Places Maximum 11 shelves
Type Shelf
Dimensions (L x W) 446 x 366 mm
Weight (Metric) 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.