missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel wire, 0.38mm (0.015in) dia, annealed, Type 304

Product Code. 11399438
Change view
Click to view available options
Quantity:
25 cm
100 cm
500 cm
Unit Size:
100cm
25cm
500cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399438 500 cm 500cm
11309448 25 cm 25cm
11319448 100 cm 100cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399438 Supplier Thermo Scientific Alfa Aesar Supplier No. 043301.CC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Temper: soft | Fe:Cr:Ni; 70:19:11 wt%

Stainless Steel Wire are suitable for making Kitchenware, Basket, Cable Trays and Trolleys with electro-polishing as the final process . Clean, de-greased & defect free surface results in very lustrous appearance when electro polished.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel
Color Silver
Name Note annealed, Type 304
Appearance Silver-colored wire
Quantity 500 cm
Odor Odorless
Form Wire
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.