Learn More
Stainless Steel wire, 0.25mm (0.01in) dia, annealed, Type 304, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 043300.BU
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Stainless Steel Wire are suitable for making Kitchenware, Basket, cable Trays and Trolleys with electro-polishing as the final process . Clean, de-greased & Defect free surface results in very lustrous appearance when electro polished.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel wire, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
25 cm | |
Odorless |
0.25mm (0.01 in.) dia., Annealed, Temper: soft | |
MFCD00148499 | |
Insoluble in water. | |
Wire |
RUO – Research Use Only