Learn More
Azpack™ Unex™ Tubing Clips
Twist nut to constrict and release tubing. Azpack™ Stainless Steel Tubing Clips are designed to distribute and apply pressure evenly.
Brand: Azpack™ 11348403
Code : 03
Additional Details : Weight : 0.05400kg
Description
- Stainless steel
- Double-banded ring for even pressure
- Nut and bolt included to apply proper compression
Specifications
Tubing Clamp | |
Tightening bolt | |
Double-banded Ring |
Stainless Steel (H10 304, A2) | |
14 to 19 mm |