missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Stainless Steel Tubing

Product Code. 13579000
Change view
Click to view available options
Overall Length (Metric):
2 m
Unit Size:
2m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) Unit Size
13579000 2 m 2m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13579000 Supplier Trajan Scientific and Medical™ Supplier No. 123751
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SilFlow is a microfluidic platform for multi-dimensional gas chromatography (GC)

  • Enhanced GC connectivity
  • Replacement ferrules and tubing available

Specifications

Type Tubing
Diameter (Metric) Outer 0.363 mm
Overall Length (Metric) 2 m
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.