missing translation for 'onlineSavingsMsg'
Learn More

Amsta Rostfritt Stainless Steel Transport Bottle Churn With Lid

Product Code. 30285050
Change view
Click to view available options
Capacity (Metric):
8 L
10 L
15 L
20 L
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30285050 8 L Pack of 10
30284863 10 L Each
30285047 15 L Each
30284967 20 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30285050 Supplier Amsta Rostfritt Supplier No. 5063281
  • ONLINE EXCLUSIVE PRICE
    £3381.38
    £2800.50 / Pack of 10
    Save £580.88
    17% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highest quality stainless steel transport bottle. Manufactured in Sweden.

  • Stainless steel 304 /1.4301

Especificaciones

Product Type Pail
Capacity (Metric) 8 L
Color Silver
Material Stainless Steel
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.