Learn More
Bochem™ Stainless Steel Tool Forceps
Product Code. 10536571
Change view
Click to view available options
Length (Metric):
160 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10536571 160 mm Each
1 options
Product Code. 10536571 Supplier Bochem™ Supplier No. 1700
Only null left
Add to basket
Add to basket
Enabling precision movement via self-tensing properties, these forceps have a sharp tip and a bent style.
Self-tensing, sharp, bent forceps. Bochem™ Stainless Steel Tool Forceps are constructed of stainless steel with a plastic handle.
  • Sharp tip
  • Bent
  • Plastic handle
  • Self-tensing
Specifications
Tip Style Sharp, Bent
Material Stainless Steel
Length (Metric) 160 mm
Product Type Forceps
Quantity (No. per Case) 10/Pack
Type Tool Forceps
Style Self-tensing