missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Test Sieves Venting Rings

Product Code. p-8004999 Shop All Retsch Products
Click to view available options
Diameter (Metric):
200 mm
203 mm
305 mm
400 mm
450 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11954422

Brand: RETSCH 69.221.3025

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Test Sieve Venting Ring
Material Stainless Steel
Diameter (Metric) 203 mm
Height (Metric) 25 mm
For Use With (Equipment) Vibratory Sieve Shaker AS 200 basic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.