missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless Steel Intermediate Pan for Test Sieves
Collects resulting samples from test sieves. These stainless steel intermediate pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.
Brand: RETSCH 69.230.0050
Additional Details : Weight : 1.20000kg
Description
- Stainless steel
- Compatible with multiple test sieves, based upon diameter
- Excellent product quality due to extensive optical inspection
- Sieve frame with a solid base
Specifications
305 mm | |
Stainless Steel | |
40mm |
Intermediate Pan | |
Test sieves; Vibratory Sieve Shaker AS 200 basic |