missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size Greater than 10 mm

Product Code. 10124782 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10124782 100 mm Each
10042822 106 mm Each
10789924 11.2 mm Each
10124732 112 mm Each
10594203 12.5 mm Each
10655743 125 mm Each
10092802 14 mm Each
10348512 13.2 mm Each
10719934 16 mm Each
10206462 18 mm Each
10544593 19 mm Each
10635933 20 mm Each
10216462 22.4 mm Each
10483412 25 mm Each
10022692 26.5 mm Each
10022702 31.5 mm Each
10493412 28 mm Each
10082722 35.5 mm Each
10266792 40 mm Each
10082732 37.5 mm Each
10092732 45 mm Each
10032762 50 mm Each
10348322 56 mm Each
10592863 53 mm Each
10695163 71 mm Each
10042762 63 mm Each
10444762 75 mm Each
10012812 80 mm Each
10144682 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
This item is not returnable. View return policy
Product Code. 10124782 Supplier RETSCH Supplier No. 60.198.100000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1 Each
Pore Size 100 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Includes Certificate indicating Compliance
Height (Metric) 50mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.