missing translation for 'onlineSavingsMsg'
Learn More

Manutan Stainless steel swivel, 1350kg break load, 6mm diameter, jaw/jaw

Product Code. 12812189
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12812189 Supplier Manutan Supplier No. A339805
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly robust swivels, providing excellent breaking strength in a compact size. Supplied in singles. Conforms to AISI 316 standard.

  • Maximum capacity: 6000kg
  • Break load: 1350kg
  • Diameter: 6mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.