missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel spheres, 1mm (0.04in) dia, Type 316

Product Code. 11395949
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395949 100 Pc. Pack of 100
11385949 25 Pc. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395949 Supplier Thermo Scientific Alfa Aesar Supplier No. 042018.KN
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel
Appearance Spheres
Quantity 100 Pc.
Form Sphere
Molecular Formula Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL Number MFCD00148499

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.