missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN55, 15 mm

Product Code. 17530286 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN55 Oven
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17530286 UN55 Oven Each
1 options
This item is not returnable. View return policy

Product Code. 17530286

Brand: Memmert™ UN55B02072

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Non-perforated slide-in drip trays constructed of stainless steel.

  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm

Specifications

For Use With (Equipment) UN55 Oven
Type Drip Tray
Material Stainless Steel
Height (Metric) Rim: 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.