missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN160, 15 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN160 Oven
Unit Size:
Each
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | UN160 Oven |
| Type | Drip Tray |
| Material | Stainless Steel |
| Height (Metric) | Rim: 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction