missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Single Tray Rack for Upright Freezers

Designed for applications where the samples are either not stored in boxes or have odd sizes. Tenak™ Single Tray Racks for Upright Freezers can be used with all freezer brands.

Brand:  Tenak™ TE29150

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 12496980

  • £88.45 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket

Description

Description

  • Trays consist of a drawer, lid, and handle on the front
  • Fits into any freezer
  • Racks are manufactured from stainless steel
Specifications

Specifications

Lid
Yes
Odd Sized Samples
1 Pack
680 x 139 x 170mm
170 mm
Single Tray Rack
Stainless Steel
Upright Freezers
1 Pack
139 mm
680 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers