Learn More
RETSCH Stainless-Steel Lid for Test Sieves
Product Code. 10781033 Shop All Retsch Products
Click to view available options
Diameter (Metric) Outer:
100 mm
150 mm
305 mm
315 mm
Unit Size:
Each
Product Code. 10781033
Brand: RETSCH 60.107.000100
Only null left
Add to basket
Add to basket
Cover sieves to avoid loss of sample during separation process. These stainless steel lids will fit multiple test sieves, based upon the sieve's diameter.
  • Stainless steel
  • Available for a variety of test sieves, based upon diameter
Specifications
Quantity 1 Ea.
Type Sieve Cover
Certifications/Compliance ISO 3310 and ASTM E 11
Cover Material Stainless Steel
Diameter (Metric) Outer 100 mm