Learn More
BINDER™ Perforated Shelf
Product Code. 17817098 Shop All BINDER Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 17817098
Brand: BINDER™ 80122175
Only null left
Add to basket
Add to basket
Material: Stainless Steel; For Use With: BF 56, FD 56, FED 56, MK 56, MKF 56
  • Perforated
  • Manufactured from stainless steel for durability and dependability
Specifications
Description Shelf, perforated - Stainless steel
For Use With (Equipment) BINDER Chambers BF 56, FD 56, FED 56, MK 56, MKF 56
Material Stainless Steel
Type Shelf