Learn More
Wheaton™ Stainless Steel Shaft Assembly Kit
Product Code. 11561295 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric) Working:
50mL
500mL
3000mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Working unitSize
11561295 500mL Each
10553252 3000mL Each
11521295 50mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11561295 Supplier Wheaton™ Supplier No. 356883
Only null left
Add to basket
Add to basket
Features an adjustable paddle blade impeller, that does not protrude through the cap, for greater turbulence
Features an adjustable paddle blade impeller, that does not protrude through the cap, for greater turbulence. The bottom dimple in 125mL and larger flasks prevents crushing of cells. 500mL and larger units have 45mm sidearms to be used as convenient vents and ports. All flasks provide a headspace ratio of 1:1 or greater to provide maximum surface interface between culture and vessel atmosphere. Unit consists of glass flask, PTFE and glass impeller assembly, and PTFE and silicone lined cap. Entire unit may be autoclaved. Optional replacement stainless steel shaft assemblies are available for each vessel. Sizes 125mL to 1,000mL are graduated in 50mL increments. 3, 000mL to 8, 000mL are graduated to 500mL increments. 25mL and 50mL are ungraduated. For microcarrier and suspension cultures such as insect cells, hybridomas and adapted cell lines.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Silicone/PTFE
Material Stainless Steel
For Use With (Application) Microcarrier and suspension cultures such as insect cells, hybridomas and adapted cell lines
Includes Glass flask, PTFE/Glass impeller assembly, and PTFE/Silicone lined cap
No. per Case 1 Each
Product Type Shaft Assembly Kit
Autoclavable Autoclavable
Closure Type Screw Cap
Volume (Metric) Working 500mL