Learn More
Vici Valco Instruments™ Stainless Steel Sample Loops
Product Code. 12757433
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12757433 100 μL Each
12747433 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12757433 Supplier Vici Valco Instruments™ Supplier No. 724JRSLR115
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems.
Vici Valco Instruments™ Stainless Steel Sample Loops are made of high-quality SS316. They are cut using an electrolytic cutting process, cleaned with ultrasound and high-pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
  • Fit all Rheodyne™ valves
  • Wide range of volumes available
  • High-pressure resistance, low permeability
Specifications
Volume (Metric) 100 μL