Learn More
Bochem™ Stainless Steel Safety Transport Containers
Product Code. 10349820
Change view
Click to view available options
Volume (Metric):
1 L
2 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10349820 2 L Each
10135600 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10349820 Supplier Bochem™ Supplier No. 8301SS
Only null left
Add to basket
Add to basket
Transport laboratory glassware safely and securely with Bochem™Stainless Steel Safety Transport Containers, which have a hinge, lock and silicone seal.
  • With hinge
  • With lock
  • With silicone seal
Specifications
Product Type Safety Transport Container
Quantity 1 Ea.
Volume (Metric) 2 L