missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel rod, 6.35mm (0.25 in.) dia., Type 303, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 013473.RM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11326838

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel rod, Type 303
Fe:Cr:Ni; 73:18:9 wt%
90 cm
6.35mm (0.25 in.) dia.
MFCD00148499
Rod
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only