Learn More
Hamilton™ PRP-X800 Cation Exchange Replacement Guard Cartridge
Product Code. 11543294
Change view
Click to view available options
:
Cation Exchange Mode, PRP-X800 Stationary Phase; Qty: 5Pack
Product Code.
11543294 Cation Exchange Mode, PRP-X800 Stationary Phase; Qty: 5Pack
1 options
Product Code. 11543294 Supplier Hamilton™ Supplier No. 79832
Only null left
Add to basket
Add to basket
A polymeric cation exchange column functionalized with itaconic acid that performs the isocratic separation of mono and divalent cations.
  • Offers excellent durability
  • Stable to any concentration organic solvent
  • Enables dynamic control of exchange capacity
  • For use with cations such as lithium, sodium, ammonium, potassium, magnesium and calcium
Specifications
Quantity 5/Pk.
Particle Size 7 μm
For Use With (Application) Cation Exchange