Learn More
Hamilton™ HxSil C18 Semiprep/Prepaarative Guard Column Replacement Cartridges
Description
- Silica packing material for a variety of reversed-phase separations
- HxSil C8 columns demonstrate excellent selectivity needed for complex sample mixtures and gives shorter retention times than a C18
- HxSil C18 columns exhibit greater retention than most columns and enhance the separation of compounds that are not sufficiently retained on other C18 columns
Specifications
Specifications
| Type | Stainless Steel Replacement Guard Cartridge. |
| Description | C18 column |