Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
Brand:  Grant Instruments™ SBT26
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15345832
  • £108.12 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
  • For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
  • Choose from a vartiety of sizes.
Specifications
Specifications
OLS26/LSB18
SBT 26 Stainless Steel Perforated Base Tray
Stainless Steel