missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel powder, -140 mesh, Type 303-L, Thermo Scientific Chemicals

Product Code. 11316098
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316098 1 kg 1kg
15450997 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316098 Supplier Thermo Scientific Alfa Aesar Supplier No. 011084.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This offers moderate corrosion resistance. Alloy 303 is frequently used in applications that require parts to be heavily machined. It is used in nuts and bolts, aircraft fittings, gears and screws.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stainless Steel powder, Type 303-L
Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:17:13 wt%
Quantity 1 kg
Mesh Size −140 Mesh
Form Powder
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.