missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate

Product Code. 10399530 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
10399530 2 g Each
10309730 100 g Each
10309540 10 g Each
10627681 20 g Each
10379820 2 kg Each
10238180 1 kg Each
10688251 200 g Each
10591911 1 g Each
10166270 500 g Each
10298080 50 g Each
10106180 5 g Each
10084140 5 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10399530 Supplier Sartorius™ Supplier No. YCW322800
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Highly polished steel calibration weights from 1g to 2kg

To OIML Class E2, F1 and F2 specification.

TRUSTED_SUSTAINABILITY

Specifications

Case Material Plastic
Mass 2 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.