missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 17-4PH, S17400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 17-4PH, S17400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Tin/Niobium/Tungsten/Nitrogen/Phosphorus/Cobalt/Vanadium/Carbon/Titanium/Molybdenum/Copper/Silicon/Manganese/Aluminium/Nickel/Chromium/Hafnium/Palladium/Yttrium/Antimony/Bismuth/Cadmium/Calcium/Lead/Selenium/Tantalum/Zinc/Zirconium/Arsenic/Oxygen/Magnesium/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0017 %/Tin 0.006 %/Niobium 0.009 %/Tungsten 0.011 %/Nitrogen 0.0153 %/Phosphorus 0.02 %/Cobalt 0.048 %/Vanadium 0.062 %/Carbon 0.08 %/Titanium 0.083 %/Molybdenum 0.35 %/Copper 0.36 %/Silicon 0.51 %/Manganese 0.73 %/Aluminium 1.09 %/Nickel 7.11 %/Chromium 17.08 %/Hafnium (<0.0010) %/Palladium (<0.0010) %/Yttrium (<0.0010) %/Antimony (<0.0050) %/Bismuth (<0.0050) %/Cadmium (<0.0050) %/Calcium (<0.0050) %/Lead (<0.0050) %/Selenium (<0.0050) %/Tantalum (<0.0050) %/Zinc (<0.0050) %/Zirconium (<0.0050) %/Arsenic (0.004) %/Oxygen (0.004) %/Magnesium (0.006) %/Iron (72.63) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction