Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Product Code. 11553264
Change view
Click to view available options
Needle Gauge:
20 G
21 G
25 G
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
11553264 25 G Pack of 6
11513264 21 G Pack of 6
11503264 20 G Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11553264 Supplier Hamilton™ Supplier No. 775116
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Autoclavable Autoclavable
Material Nickel-plated Brass (Hub), Stainless Steel
Needle Gauge 25 G
Diameter (Metric) Inner 0.26 mm
Diameter (Metric) Outer 0.52 mm
Product Type Luer Lock Custom Needle
Quantity 6/Pk.
Needle Point Style 5 or AS
For Use With (Equipment) TLL Syringes and LT or TLL Connectors
Connection Type Standard Tapered