Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 5 or AS
Product Code. 11503264
Click to view available options
Needle Gauge:
20 G
21 G
25 G
Unit Size:
Pack of 6
Product Code. 11503264
Brand: Hamilton™ 775111
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Material Nickel-plated Brass (Hub), Stainless Steel
Needle Gauge 20 G
Diameter (Metric) Inner 0.60 mm
Diameter (Metric) Outer 0.91 mm
Product Type Luer Lock Custom Needle
Quantity 6/Pk.
Needle Point Style 5 or AS
For Use With (Equipment) TLL Syringes and LT or TLL Connectors
Connection Type Standard Tapered